Learn More
Sino Biological Recombinant Human Fetuin A/FETUA Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10318H08H100
Encompass_Preferred
A DNA sequence encoding the human Fetuin A (NP_001613.2) (Met 1-Val 367) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4586