Learn More
Sino Biological Anti-Fetuin A/FETUA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10318T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Fetuin-A (rh Fetuin-A; Catalog#10318-H08H; NP_001613.2; Met1-Val367). Fetuin-A specific IgG was purified by Human Fetuin-A affinity chromatography.
Catalog No. 50-172-4431