Learn More
Sino Biological Recombinant Human Leptin Receptor Protein (His & hFc Tag), HPLC-verified
Supplier:  Sino Biological 10322H03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Asp 839) of human leptin receptor precursor (NP_002294.2) was expressed with the C-terminal fused polyhistidine-tagged Fc region of human IgG1.
Catalog No. 50-161-4588