Learn More
Sino Biological Anti-Leptin Receptor Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10322MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Leptin Receptor / CD295 / LEPR (rh Leptin Receptor / CD295 / LEPR; Catalog#10322-H08H; NP_002294.2; Met 1-Asp 839). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4457