Learn More
Sino Biological Anti-Leptin Receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10322RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Leptin Receptor / CD295 / LEPR (rh Leptin Receptor / CD295 / LEPR; Catalog#10322-H08H; NP_002294.2; Met 1-Asp 839). Leptin Receptor / CD295 / LEPR specific IgG was purified by Human Leptin Receptor / CD295 / LEPR affinity chromatography.
Catalog No. 50-172-4467