Learn More
Sino Biological Anti-HAI-2/SPINT2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10324T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SPINT2 (rh SPINT2; Catalog#10324-H08H; NP_066925.1; Met1-Lys197). SPINT2 specific IgG was purified by Human SPINT2 affinity chromatography.
Catalog No. 50-172-4479