Learn More
Sino Biological Recombinant Human HB-EGF Protein
Supplier:  Sino Biological 10325HNAB10
Encompass_Preferred
A DNA sequence encoding the human HBEGF (Q99075) (Met 1-Leu 148) without the C-terminal propeptide (aa 149-208), was expressed and purified.
Catalog No. 50-161-4598