Learn More
Sino Biological Recombinant Human HMGB1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10326H07E50
Encompass_Preferred
A DNA sequence encoding the mature form of human HMGB1 (P09429) (Lys57-Asp158) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-4604