Learn More
Sino Biological Recombinant Human HMGB1 Protein (His Tag), HPLC-verified
Catalog No. 501614605
Encompass_Preferred
A DNA sequence encoding the human HMGB1 protein (NP_002119.1) (Met 1-Glu 215) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-161-4605 Supplier Sino Biological Supplier No. 10326H08H100
A DNA sequence encoding the human HMGB1 protein (NP_002119.1) (Met 1-Glu 215) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.