Learn More
Sino Biological Recombinant Human MMP-9 Protein, HPLC-verified
Supplier:  Sino Biological 10327HNAH20
Encompass_Preferred
A DNA sequence encoding the human MMP9 zymogen (NP_004985.2) (Met 1-Asp 707) was expressed.
Catalog No. 50-161-4610