Learn More
Sino Biological Recombinant Anti-MMP-9 Antibody (Biotin), Rabbit Monoclonal
Supplier:  Sino Biological 10327R043B100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human matrix metallopeptidase 9 (rh MMP-9; Catalog#10327-H08H; Met 1-Asp 707; NP_004985.2) and then biotinylated.
Catalog No. 50-172-4504