Learn More
Sino Biological Anti-HGF Activator/HGFA Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10329MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human HGFA (rhHGFA; Catalog#10329-H08H; NP_001519.1; Met 1-Ser 655). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4519