Learn More
Sino Biological Anti-HGF Activator/HGFA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10329RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HGFA (rhHGFA; Catalog#10329-H08H; NP_001519.1; Met 1-Ser 655). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-4524