Learn More
Sino Biological Recombinant Human GFR Alpha-1/GFRA1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10330H08H100
Encompass_Preferred
A DNA sequence encoding the human GFRα1 without the propeptide (NP_665736.1) (Met 1-Ser 424) was fused with the a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4615