Learn More
Sino Biological Recombinant Human ICAM-2/CD102 Protein (His Tag)
Supplier:  Sino Biological 10332H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 223) of human ICAM2 (NP_000864.2) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4621