Learn More
Sino Biological Anti-ICAM-2/CD102 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10332MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ICAM-2 / CD102 (rh ICAM-2 / CD102; Catalog#10332-H08H; NP_000864.2; Met 1-Gln 223). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4543