Learn More
Sino Biological Recombinant Anti-ICAM-2/CD102 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10332R015100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ICAM-2/CD102 (rh ICAM-2/CD102; Catalog#10332-H08H; NP_000864.2; Met1-Gln223).
Catalog No. 50-172-4550