Learn More
Sino Biological Recombinant Anti-ICAM-2/CD102 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10332R041100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ICAM2 (rh ICAM2; Catalog#10332-H08H; NP_000864.2; Met 1-Gln 223).
Catalog No. 50-172-4557