Learn More
Sino Biological Anti-ICAM-2/CD102 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10332RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ICAM-2 / CD102 (rh ICAM-2 / CD102; Catalog#10332-H08H; NP_000864.2; Met 1-Gln 223). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-4561