Learn More
Sino Biological Recombinant Human ICAM-3/CD50 Protein (His & hFc Tag)
Supplier:  Sino Biological 10333H03H100
Encompass_Preferred
The extracellular domain (Met 1-His 485) of human ICAM3 (NP_002153.2) precursor was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4623