Learn More
Sino Biological Recombinant Human ICAM-3/CD50 Protein (His Tag)
Supplier:  Sino Biological 10333H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human ICAM3 (CAA49369.1) (Met 1-His 485) was expressed, with the C-terminal fused polyhistidine tag.
Catalog No. 50-161-4625