Learn More
Sino Biological Anti-ICAM-3/CD50 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10333MM02F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD50 / ICAM-3 (rh CD50 / ICAM-3; Catalog#10333-H08H; CAA49369.1; Met 1-His 485) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-4577