Learn More
Sino Biological Anti-ICAM-3/CD50 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10333MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ICAM3 (rh ICAM3; Catalog#10333-H08H; CAA49369.1; Met 1-His 485). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4580