Learn More
Sino Biological Recombinant Anti-ICAM-3/CD50 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10333R101100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD50 / ICAM-3 (rh CD50 / ICAM-3; Catalog#10333-H08H; CAA49369.1; Met 1-His 485).
Catalog No. 50-172-4587