Learn More
Sino Biological Recombinant Human HVEM Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10334H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Val202) of human TNFRSF14 (NP_003811.2) was expressed fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-4631