Learn More
Sino Biological Anti-HVEM Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10334MM11P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human HVEM/TNFRSF14/CD270 (rh HVEM/TNFRSF14/CD270; Catalog#10334-H08H; NP_003811.2; Met1-Val202) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-4601