Learn More
Sino Biological Recombinant Anti-HVEM Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10334R004A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human HVEM/TNFRSF14/CD270 (rh HVEM/TNFRSF14/CD270; Catalog#10334-H08H; NP_003811.2; Met1-Val202) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-4608