Learn More
Sino Biological Anti-E-Selectin/CD62E Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10335MM10P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human E-Selectin / CD62e (rh E-Selectin / CD62e; Catalog#10335-H08H; NP_000441.2; Met 1-Pro 556) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-4638