Learn More
Sino Biological Recombinant Anti-E-Selectin/CD62E Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10335R103F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human E-Selectin / CD62e (rh E-Selectin / CD62e; Catalog#10335-H08H; NP_000441.2; Met 1-Pro 556) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-4647