Learn More
Sino Biological Recombinant Human Interferon alpha 4/IFNA4 Protein (hFc Tag)
Supplier:  Sino Biological 10336H01H100
Encompass_Preferred
A DNA sequence encoding the mature form of human IFN-α4a (NP_066546.1) (Cys 24-Asp 189) was expressed with the N-terminal fused Fc region of human IgG1.
Catalog No. 50-161-4637