Learn More
Sino Biological Anti-Interferon alpha 4/IFNA4 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10336MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IFNA4 (rh IFNA4; Catalog#10336-H08B; NP_066546.1; Met1-Asp189). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4662