Learn More
Sino Biological Recombinant Anti-Interferon alpha 4/IFNA4 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10336R053100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IFNA4 (rh IFNA4; Catalog#10336-H08B; NP_066546.1; Met1-Asp189).
Catalog No. 50-172-4665