Learn More
Sino Biological Anti-Interferon alpha 4/IFNA4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10336RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IFNA4 (rh IFNA4; Catalog#10336-H08B; NP_066546.1; Met1-Asp189). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-4667