Learn More
Sino Biological Recombinant Human IFNGR1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10338H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gly 245) of human IFN-γR1 pre-protein (NP_000407.1) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4649