Learn More
Sino Biological Anti-IFNGR1 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10338MM05P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IFNGR1 / CD119 (rh IFNGR1 / CD119; Catalog#10338-H08H; NP_000407.1; Met1-Gly245) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-4688