Learn More
Sino Biological Recombinant Anti-IFNGR1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10338R010100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IFNGR1 / CD119 (rh IFNGR1; Catalog#10338-H08H; NP_000407.1; Met 1-Gly 245).
Catalog No. 50-172-4692