Learn More
Sino Biological Anti-SMAC Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10339MM12100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SMAC / Diablo (rh SMAC / Diablo ; Catalog#10339-H08E; NP_063940.1; Ala56-Asp239). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4703