Learn More
Sino Biological Anti-SMAC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10339RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Diablo (rh Diablo; Catalog#10339-H08E; NP_063940.1; Ala 56-Asp 239). Diablo specific IgG was purified by human Diablo affinity chromatography.
Catalog No. 50-172-4717