Learn More
Sino Biological Recombinant Human SLITRK1 Protein (His & hFc Tag)
Supplier:  Sino Biological 10340H03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ser 616) of human SLITRK1 (NP_443142.1) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4653