Learn More
Sino Biological Recombinant Human SLITRK1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10340H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ser 616) of human SLITRK1 (NP_443142.1) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4655