Learn More
Sino Biological Anti-SLITRK1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10340RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SLITRK1 (rh SLITRK1; Catalog#10340-H08H; NP_443142.1; Met 1-Ser 616). SLITRK1 specific IgG was purified by human SLITRK1 affinity chromatography.
Catalog No. 50-172-4728