Learn More
Sino Biological Anti-PACSIN1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 103403T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PACSIN1, and purified by antigen affinity chromatography.
Catalog No. 50-172-4720