Learn More
Selleck Chemical LLC Disodium (R)-2-Hydroxyglutarate 25mg 103404-90-6
SDP
Supplier:  Selleck Chemical LLC S787325MG
Encompass_Preferred
Disodium (R)-2-Hydroxyglutarate is a competitive inhibitor of ?-ketoglutarate-dependent dioxygenases with Ki of 10.87 ¿ 1.85 mM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6400