Learn More
Sino Biological Recombinant Human interferon-alpha/IFNA5 Protein (hFc Tag)
Supplier:  Sino Biological 10342H01H100
Encompass_Preferred
A DNA sequence encoding the human IFNα G (NP_002160.1) (Leu 22-Glu 189) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4662