Learn More
Sino Biological Recombinant Human interferon-alpha/IFNA5 Protein (His Tag)
Supplier:  Sino Biological 10342H08Y100
Encompass_Preferred
A DNA sequence encoding the human IFNA5 (NP_002160.1) (Leu22-Glu189) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4665