Learn More
Sino Biological Anti-interferon-alpha/IFNA5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10342T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IFNA5 / IFNaG (rh IFNA5 / IFNaG; Catalog#10342-H08H; P01569; Met1-Glu189). IFNA5 / IFNaG specific IgG was purified by Human IFNA5 / IFNaG affinity chromatography.
Catalog No. 50-172-4742