Learn More
Sino Biological Anti-AIF1L Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 103429T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human AIF1L, and purified by antigen affinity chromatography.
Catalog No. 50-172-4731