Learn More
Sino Biological Recombinant Human SLPI Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10343H08B100
Encompass_Preferred
A DNA sequence encoding the human SLPI (P03973)( Met1-Ala132) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-4669