Learn More
Sino Biological Anti-SLPI Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10343MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SLPI (rh SLPI; Catalog#10343-H08B; P03973; Met1-Ala132). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4757