Learn More
Sino Biological Anti-ALDH3A2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 103433T44100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse ALDH3A2, and purified by antigen affinity chromatography.
Catalog No. 50-172-4745